SpillStopper™ Work Surfaces for Protector® ClassMate™ Laboratory Hoods, Labconco®
Supplier: Labconco
9600402 9600410 9600420 9600411 9600501 9600502 9600510 9600520 9600511 9600522 9600512 9600521 9600620 9600611 9600622 9600612 9600621 9600400 9600401 9600422 9600412 9600421 9600500 9600600 9600601 9600602 9600610
76045-658EA 4558.3 CAD
76045-658 76045-660 76045-662 76045-664 76045-666 76045-668 76045-670 76045-672 76045-674 76045-676 76045-678 76045-680 76045-682 76045-684 76045-686 76045-688 76045-690 76045-924 76045-926 76045-928 76045-930 76045-932 76045-934 76045-936 76045-938 76045-940 76045-942
SpillStopper™ Work Surfaces for Protector® ClassMate™ Laboratory Hoods, Labconco®
Fume Hoods and Enclosures Chemical Fume Hoods
Solid epoxy work surfaces are 3.2cm (11/4") thick, 65.7cm (257/8") deep, and dished to contain spills.
Solid epoxy cupsink cover (model 4005201; sold separately) covers cupsink when not in use.