VWR® Qualitative Filter Paper, Grade 410
Supplier: VWR International
28297-826 28321-077 28321-113 28321-135 CA28321-077 CA28297-826 CA28321-113 CA28321-135
28297-826PK 70.23 CAD
28297-826 28321-077 28321-113 28321-135 CA28321-077 CA28297-826 CA28321-113 CA28321-135
VWR® Qualitative Filter Paper, Grade 410
Filters Filter Papers Qualitative Filter Paper
Grade 410 filter paper is useful in analytical operations involving filtration of cuprous oxide, lead sulfate, barium sulfate, manganese carbonate, and more.