VWR® Quantitative Filter Paper, Grade 454
Supplier: VWR International
28306-051 28306-095 28306-110 28306-131 28306-153 28306-175 28306-051 28306-095 28306-110 28306-131 28306-153 28306-175
28306-051PK 42.36 CAD
28306-051 28306-095 28306-110 28306-131 28306-153 28306-175 CA28306-051 CA28306-095 CA28306-110 CA28306-131 CA28306-153 CA28306-175
VWR® Quantitative Filter Paper, Grade 454
Filters Filter Papers Quantitative Filter Paper
Use for retention of zirconium hydroxide, aluminum hydroxide, cobalt sulfide, and other coarse or gelatinous precipitates.