StatSpin® Microhematocrit Tubes, Iris Sample Processing
Supplier: Statspin
HP8H-10 HP4H-10 HP4U-10 HT9H-10 HT9U-10 HP8U-10
CA56430-046CS 557.75 CAD
CA56430-046 CA56430-048 CA56430-050 CA56430-052 CA56430-054 CA56430-084
StatSpin® Microhematocrit Tubes, Iris Sample Processing
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
These microhematocrit tubes use capillary action to draw samples quickly.
  • Capillary Action Draws Samples Quickly
  • Material: Non-Breakable Plastic for Safety
  • Heparin Tubes for Finger-Stick Samples
  • Untreated Tubes for Anti-Coagulated Venous Samples
  • Available Lengths: 75 mm & 40 mm
Tubes are available with heparin for finger-stick samples or untreated for anti-coagulated venous samples
SafeCrit® hematocrit tubes are made of non-breakable, 100% plastic to eliminate the potential safety hazards of broken glass.
The 40 mm tubes are made for use with the MP multipurpose centrifuge (87002-248 series) and the CritSpin™ microhematocrit centrifuge (87002-206 series). The 75 mm tubes are made for use in conventional centrifuges that support the full length of the tube.