Eppendorf® 5804 R Refrigerated Benchtop Centrifuge

Supplier: Eppendorf
022623508 022625080 022628146 022628045 022628153 022628051 022628236 022628225
CA53513-804EA 19969.68 CAD
CA53513-804 CA53513-806 CA80076-422 CA89210-974 CA89210-976 CA11017-352 CA11017-356 490013-048 CA89210-978 76460-826
Eppendorf® 5804 R Refrigerated Benchtop Centrifuge
Centrifuges Benchtop Centrifuges
The high-speed refrigerated Eppendorf Centrifuge 5804 R is a medium capacity centrifuge, ideal for molecular applications in tubes up to 250 ml. It also offers supreme versatility with the capability of taking swing-bucket and fixed-angle rotors, as well as deepwell plates.

  • Low access height of 29 cm for easy loading and unloading of samples
  • Quiet operation to improve work environment
  • Compact footprint saves valuable bench space
  • Automatic rotor recognition and imbalance detection for maximum operational safety
  • Swing-bucket rotors and adapters accommodate tubes and bottles from 0.2 to 250 ml
  • Plate rotor for centrifugation of all types of MTP, PCR, cell culture or deepwell plates
  • Fixed-angle rotors for high-speed applications in tubes from 0.2 to 85 ml
  • FastTemp function for fast pre-cooling
  • Continuous cooling maintains a constant temperature after the run has been completed - samples stay cool
  • ECO shut-off engages after 8 hours of non-use to reduce energy consumption and to extend compressor life
  • Dynamic compressor control (DCC) technology for optimized cooling performance

Centrifuge 5804 R, a refrigerated benchtop centrifuge renowned for its quality and reliability, offers the most cost-effective solution for medium-throughput applications requiring speeds of up to 20,913×g (14,000 rpm). Experience true versatility with a broad selection of rotors and adapters that accommodate tubes and bottles from 0.2 to 250 ml, as well as MTP, PCR and deep-well plates.

The swing-bucket rotor and adapters accommodate tubes and bottles from 0.2 to 250 ml whereas the fixed-angle rotors accomodate vessels from 0.2 to 85 ml. The plate rotor is ready to carry out centrifugation of all common types of multiwell plate, such as PCR, cell culture or deep-well plates.

Max. Capacity Plates: 10×MTP/2×DWP
Rotors Available: 12

Certifications: UL Listed, Certificate of Quality/Conformity (ISO 9001, ISO 13485, ISO 14001).
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR