Whatman™ Multiple Vacuum Filtration Apparatus, Three and Six-Place Vacuum Filtration System, Whatman products (Cytiva)
Supplier: Cytiva
10445835 10444835 10498762 10444835 10445835 10498762 10498761 10498761
CA28151-450EA 8616.63 CAD
CA28151-450 CA28151-456 CA28151-458 CA28151-460 CA28151-466 CA28151-468 CA97048-734 28151-450 28151-460 28151-468 28151-456 28151-458 28151-466 97048-734 89233-738 CA89233-738
Whatman™ Multiple Vacuum Filtration Apparatus, Three and Six-Place Vacuum Filtration System, Whatman products (Cytiva)
Manifolds
Multiple vacuum filtration apparatus suitable only for vacuum operation.
  • For microbiological quality control
  • Residue analyses
  • Serial filtration carried out rapidly and easily with a common drainage outlet
Stainless steel including sieve filter support
Individual stopcocks, 47/50 mm filter size, tubing nozzle 9 mm vacuum connection.
Caution: Suitable only for vacuum. Do not exceed 1.3bar (300mbar overpressure) when using flushing tubes.