Stainless Steel Hand Drying Wipe Dispensers
Supplier: Contec
9451DW 9451DWS 9451
77705-708EA 304.15 CAD
77705-708 77705-710 77705-706
Stainless Steel Hand Drying Wipe Dispensers
Wipe Dispensers
Hand drying wipe dispensers are available in two wall-mounted formats (adhesive backed or screw thread) or as a floor-standing unit.
  • Stainless steel construction
  • Designed for use with large, absorbent 43×23cm (17×9") low-lint hand drying wipes
  • Durable and corrosion-resistant for long-lasting use in cleanrooms and controlled environments
  • Other dispensing alternatives can be open to the environment and do not protect wipes from contamination
The wall-mounted hand dry wipe dispensers are available in two mounting options. Dispensers with adhesive backing offer a secure, space-saving solution for easy wipe access. The strong adhesive ensures stability on various surfaces. The hand dry wipe dispenser with screw mount offers secure and durable mounting to ensure long-term stability on walls or surfaces.
The floor-standing hand wipe dispenser offers a durable, easy-to-clean solution for efficient wipe dispensing. This model features an optimized height and stable base for easy and comfortable dispensing. It is also easy to relocate as needed.
All models ensure reliability and hygiene and are ideal for use in cleanrooms and controlled environments.