Mouse Recombinant Rank Ligand (from E. coli)
Supplier: VWR International
77751-920
77751-922
77751-924
77751-926
77751-920EA
1464.15
CAD
77751-920
77751-922
77751-924
77751-926
Mouse Recombinant Rank Ligand (from E. coli)
Proteins and Peptides
Receptor activator of nuclear factor kappa-B Ligand (RANK Ligand) is a cell-bound marker related to the tumor necrosis factor (TNF) family of proteins. RANK Ligand plays a critical role in bone metabolism and osteoclast differentiation. T cell expression of RANK Ligand promotes dendritic cell maturation.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.