Safety Caps for GL 45 Threads
Supplier: CP Lab Safety
ALW-GL45-2KIT ALW-GL45-3KIT ALW-GL45-4KIT
77676-556EA 93.22 CAD
77676-556 77676-558 77676-560
Safety Caps for GL 45 Threads
Closures Safety Caps and Safety Waste Caps
2-Port safety cap deluxe kit, GL45 thread, black outer, ready-to-use with air inlet valve and and ¹/₈" OD tubing adapter. All you need to supply is the ¹/₈" OD tubing and you're ready to work! Create a closed system with GL-45 safety port caps, which seal to a bottle with a secure EPDM O-ring resistant to common HPLC solvents. This cap helps avoid twisted tubing due to its free-spinning collar design. The inner PTFE section will not rotate after the cap is secured. For a closed system, use the adapter with ¹/₈" OD tubing to seal one port and a vent filter in the other to allow for pressure changes and off-gassing.
  • Includes plugs for unused ports
  • Use ¹/₄-28 adapters to fit ¹/₈ or ¹/₁₆" OD tubing (not included) which can fit through the holes and into the bottom of the bottle
  • Fits any GL45 media bottle
  • ideal for solvent delivery applications such as HPLC
  • prevents evaporation of volatile compounds
  • polypropylene outer and PTFE inner
  • avoid twisted tubing with free-spinning collar
3-Port solvent delivery port cap kit for GL45 media bottles, red, with plugs, for use with ¹/₈ or ¹/₁₆" OD tubing in ¹/₄-28 adapters. Create a closed system with GL-45 safety caps, which seal to a bottle with a secure EPDM O-ring resistant to common HPLC solvents. All you need to supply is your tubing and the ¹/₄-28 UNF threaded adapters of your choice.
4-Port solvent delivery port cap kit for GL45 media bottles, yellow, with plugs, for use with ¹/₈ or ¹/₁₆" OD tubing in ¹/₄-28 adapters. Create a closed system with GL-45 safety caps, which seal to a bottle with a secure EPDM O-ring resistant to common HPLC solvents. All you need to supply is your tubing and the ¹/₄-28 UNF threaded adapters of your choice. Note: We suggest also using an optional vent filter to allow for pressure changes and off-gassing.