CleanZorb® Cleanroom Nonwoven Wipers
Supplier: Connecticut Cleanroom
CR12-150 CR18-150 CR4-1200 CR7-300 CR9-150
77687-016EA 40.34 CAD
77687-016 77687-018 77687-020 77687-024 77687-026
CleanZorb® Cleanroom Nonwoven Wipers
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Ideal for applications where high sorbency and low extractables are important. Excellent for picking up aqueous spills. Suitable for cleaning precision components and apparatus.
  • CleanZorb® Nonwoven, spunlaced construction for excellent strength
  • Compatible with most solvents
  • Can be gamma irradiated
  • Excellent chemical resistance for compatibility with a variety of solutions
  • Autoclave safe (dry wipers only)
Fabricated from a blend of 45% polyester and 55% cellulose, this material combines the high sorbency of natural fibers with the cleanliness of synthetic fibers. Low particle counts, low extractable levels and solvent compatibility, this wiper is ideal for general cleaning, wiping and spill control in controlled environments.
Applications:
Wiping and cleaning surfaces, equipment and parts.
Spill control.
Applying and removing lubricants, adhesives, residues, other solutions including disinfectants.
Cleaning with solvents such as isopropyl alcohol (IPA), ethanol, acetone, and degreasers.
Used in industries such as Aerospace (compliant with SS9170/680), Animal laboratory, Biologics, Cleanroom design/build, Compounding, Pharmacies data storage, Facilities, Maintenance industrial laboratory, Medical device, Microelectronics, Pharmaceutical, Printing/graphics, Semi-conductors.
Certifications: Meets USP <797> and <800> wiper requirements. Cleanroom Environment ISO Class 5 - 8, Class 100 - 100000, EU Grade B - D.