SafeBasics™ Self-Sealing Sterilization Pouches
Supplier: Medicom
99000 99005 99010 99010B 99015 99025 99030
CA77749-870BX 16.43 CAD
CA77749-870 CA77749-872 CA77749-874 CA77749-876 CA77749-878 CA77749-880 CA77749-882
SafeBasics™ Self-Sealing Sterilization Pouches
Bags Sterilization Pouches
Provides an effective method to enclose devices (skin and mucosal tissue contact) intended for sterilization in steam autoclaves and via Ethylene Oxide (EO). Dual process indicators designed to indicate to the user that the pouch has undergone either a steam or EO sterilization process.
  • Reliable indicator color change and seal performance
  • Triple seal with larger width for increased seal strength
  • Internal multi-variable indicators confirm the proper sterilization parameters inside the pouch has been reached
  • External process indicators distinguish between processed and unprocessed contents
  • Non-adhesive edge allows for easy peel and airtight seal, with or without gloves
  • Green film provides good visibility of contents, including color-coded instruments, and indicator color, as well as pinholes, tears, or other damage
  • Internal isolating seal keeps instruments from interfering with the internal, multi-parameter chemical indicator
  • Thumb loop notch facilitates a clean aseptic opening of the pouch post sterilization
Made from high quality materials, Medicom SafeBasics™ pouches are designed to ensure successful sterilization processing you can trust. Efficiently packaged in compact boxes to help reduce carbon footprint.
Green tinted film sealed to paper with two sets of indicators and double sides coated adhesive tape.
Steam: Indicators turn from blue to black when processed 30 min at 121 °C.
EO: Indicators turn from pink to brown when processed 40 min at 60 °C, 60% RH, and EO concentration of 725 mg/L.