SafeSeal® Quattro Self-Sealing Sterilization Pouches
Supplier: Medicom
88045-4 88000-4 88005-4 88010-4 88010-4B 88015-4 88020-4 88025-4 88030-4 88035-4 88040-4
CA77749-868BX 62.81 CAD
CA77749-868 CA77749-848 CA77749-850 CA77749-852 CA77749-854 CA77749-856 CA77749-858 CA77749-860 CA77749-862 CA77749-864 CA77749-866
SafeSeal® Quattro Self-Sealing Sterilization Pouches
Bags Sterilization Pouches
Intended for sterilization in steam autoclaves or via Ethylene Oxide (EO). The pouch's process indicators are designed to indicate to the user that the pouch has undergone either a steam or EO sterilization process.
  • Medical grade paper and European-sourced film helps reduce curling and the risk of punctures and tears
  • Internal multi-parameter and external process indicators reacts to time, temperature, and pressure
  • Thumb loop notch facilitates a clean aseptic opening of the pouch post sterilization
  • Extra wide chevron with TruePress™ Technology seal protects against instrument breaching, delamination, and ensures a clean separation
  • Wide self-seal adhesive strip with perforated fold facilitates accuracy in forming an airtight seal
Made with high quality materials in our state-of-the-art facility in North America, Medicom Quattro pouches ensure reliable sterilization so you can practice with confidence.
Blue tinted film sealed to paper with two sets of indicators and double sides coated adhesive tape.
Steam: Indicators turn from pink to cocoa when processed 30 min. at 121 °C or 4 min. at 134 °C.
EO: Indicators turn from blue to golden brown when processed 100 to 120 min at 50 °C, 60 t 85% RH, and EO concentration of 600 mg/L.