Anti-Thymocyte globulin Rabbit polyclonal Antibody

Supplier: Creative Biomart
FMD-458-1MG FMD-458-5MG
77682-304EA 1561.07 CAD
77682-304 77682-302
Anti-Thymocyte globulin Rabbit polyclonal Antibody
Antibodies
Thymoglobulin is a polyclonal antibody that suppresses certain types of immune cells responsible for acute organ rejection in transplant patients. Thymoglobulin is a mixture of antibodies intended to bind to various cell surface antigens. The most common mode of action of Thymoglobulin is via selective depletion of T-cells.

Type:
Antigen: Thymocyte globulin
Clonality: polyclonal 
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR