Anti-DEFB1 Rabbit Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A43841-50UG A43841-100UG
77706-606EA 360.84 CAD
77706-606 77706-605
Anti-DEFB1 Rabbit Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: DEFB1
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR