Anti-CMTM2 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A43817-100UL A43817-50UL
77706-562EA 529.22 CAD
77706-562 77706-563
Anti-CMTM2 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:10000, IHC:1:100-1:300

Type:
Antigen: CMTM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR