Anti-CALCA Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A43716-50UL A43716-100UL
77706-381EA 360.84 CAD
77706-381 77706-380
Anti-CALCA Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:5000, IHC:1:50-1:200

Type:
Antigen: CALCA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR