Anti-DEFA1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A43676-100UL A43676-50UL
77706-317EA 529.22 CAD
77706-317 77706-318
Anti-DEFA1 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:500-1:5000, IHC:1:5-1:20

Type:
Antigen: DEFA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Order Now

SPECIFICATIONS

Antigen symbol DEFA1
Environmentally Preferable
Antigen synonyms DEF1_HUMAN|DEFA1B|MRS|Neutrophil defensin 2|alpha 1|HP-1|HNP1|HP2|HP-2|alpha 2|HNP-2|HNP-1|Defensin 1|DEF1|HP1|Defensin|Neutrophil defensin 1|Myeloid related sequence|DEFA2|DEFA1
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Gene ID P59665
Isotype IgG
ELISA Yes
ImmunoChemistry Yes
Immunogen Synthetic peptide of Human DEFA1
Form Liquid
Purification Antigen affinity purified
Storage buffer PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
Storage temperature Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
Shipping temperature Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR