Anti-ARHGEF5 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A43564-50UL A43564-100UL
77706-119EA 360.84 CAD
77706-119 77706-118
Anti-ARHGEF5 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:10000, WB:1:1000-1:5000, IHC:1:50-1:200

Type:
Antigen: ARHGEF5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR