Anti-EEF2 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A43554-50UL A43554-100UL
77706-101EA 360.84 CAD
77706-101 77706-100
Anti-EEF2 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:2000, WB:1:500-1:2000, IHC:1:25-1:100

Type:
Antigen: EEF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR