Anti-ATP2A3 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A43476-50UL A43476-100UL
77705-958EA 360.84 CAD
77705-958 77705-957
Anti-ATP2A3 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:2000, IHC:1:25-1:100

Type:
Antigen: ATP2A3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR