Anti-CABLES2 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A43402-50UL A43402-100UL
77705-836EA 360.84 CAD
77705-836 77705-835
Anti-CABLES2 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:2000, IHC:1:25-1:100

Type:
Antigen: CABLES2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR