Anti-Nsf Rabbit Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A46056-100UG A46056-50UG
77710-515EA 529.22 CAD
77710-515 77710-516
Anti-Nsf Rabbit Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: Nsf
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR