Anti-Nsf Rabbit Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A46055-100UG A46055-50UG
77710-513EA 529.22 CAD
77710-513 77710-514
Anti-Nsf Rabbit Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: Nsf
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR