Anti-ACE Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A45976-50UL A45976-100UL
77710-383EA 360.84 CAD
77710-383 77710-382
Anti-ACE Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:10000, WB:1:1000-1:5000

Type:
Antigen: ACE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR