Anti-AIF1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A45896-50UL A45896-100UL
77710-243EA 360.84 CAD
77710-243 77710-242
Anti-AIF1 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:2000, WB:1:200-1:1000, IHC:1:50-1:200

Type:
Antigen: AIF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR