Anti-TFR2 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A45788-50UG A45788-100UG
77710-055EA 360.84 CAD
77710-055 77710-054
Anti-TFR2 Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: TFR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR