Anti-STX12 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A45683-50UL A45683-100UL
77709-868EA 360.84 CAD
77709-868 77709-867
Anti-STX12 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:10000, WB:1:1000-1:5000

Type:
Antigen: STX12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR