Anti-RUNX2 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A45672-50UG A45672-100UG
77709-848EA 360.84 CAD
77709-848 77709-847
Anti-RUNX2 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:500-1:5000

Type:
Antigen: RUNX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Order Now

SPECIFICATIONS

Environmentally Preferable
Antigen synonyms Acute myeloid leukemia 3 protein|PEA2aA|CBF alpha 1|PEBP2aA1|CBFA1|PEBP2-alpha A|SL3/AKV core-binding factor alpha A subunit|Core binding factor runt domain alpha subunit 1|Polyomavirus enhancer-binding protein 2 alpha A subunit|OSF2|OSF-2|CCD1|Oncogene AML 3|PEBP2aA|Core-binding factor subunit alpha-1|RUNX2|OTTHUMP00000016533|Polyomavirus enhancer binding protein 2 alpha A subunit|Runt-related transcription factor 2|SL3/AKV core binding factor alpha A subunit|Core binding factor|RUNX2_HUMAN|PEA2 alpha A|PEBP2A2|MGC120022|PEA2-alpha A|Osteoblast specific transcription factor 2|SL3 3 enhancer factor 1 alpha A subunit|Runt related transcription factor 2|Cleidocranial dysplasia 1|CBF-alpha-1|Osteoblast-specific transcription factor 2|Runt domain|MGC120023|OSF 2|CCD|SL3-3 enhancer factor 1 alpha A subunit|AML3|Core binding factor subunit alpha 1|Alpha subunit 1|Oncogene AML-3|PEBP2A1|PEBP2 alpha A
Reactivity
Host
Gene ID Q13950
Isotype
Immunogen Recombinant Human Runt-related transcription factor 2 protein (219-404AA)
Form Liquid
Purification >95%, Protein G purified
Storage buffer 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
Storage temperature Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
Shipping temperature Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR