Anti-NANOS2 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A45578-100UL A45578-50UL
77709-686EA 529.22 CAD
77709-686 77709-687
Anti-NANOS2 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, IHC:1:50-1:200

Type:
Antigen: NANOS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR