Anti-HMMR Rabbit Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A45576-50UG A45576-100UG
77709-683EA 360.84 CAD
77709-683 77709-682
Anti-HMMR Rabbit Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: HMMR
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR