Anti-ADAMTS15 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A45487-100UL A45487-50UL
77709-529EA 529.22 CAD
77709-529 77709-530
Anti-ADAMTS15 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:2000, WB:1:200-1:1000

Type:
Antigen: ADAMTS15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR