Anti-RNF7 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A45372-100UL A45372-50UL
77709-327EA 529.22 CAD
77709-327 77709-328
Anti-RNF7 Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: RNF7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR