Anti-MTFP1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A45263-50UL A45263-100UL
77709-129EA 360.84 CAD
77709-129 77709-128
Anti-MTFP1 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, WB:1:500-1:2000, IHC:1:25-1:100

Type:
Antigen: MTFP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR