Anti-IFNL1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A45250-100UG A45250-50UG
77709-106EA 529.22 CAD
77709-106 77709-107
Anti-IFNL1 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: IHC:1:20-1:200

Type:
Antigen: IFNL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR