Anti-DUSP8 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A45247-100UL A45247-50UL
77709-101EA 529.22 CAD
77709-101 77709-102
Anti-DUSP8 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:10000, IHC:1:50-1:200

Type:
Antigen: DUSP8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR