Anti-CANT1 Rabbit Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A45193-100UG A45193-50UG
77709-003EA 529.22 CAD
77709-003 77709-004
Anti-CANT1 Rabbit Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: CANT1
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR