Anti-CIDEB Rabbit Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A45115-50UG A45115-100UG
77708-863EA 360.84 CAD
77708-863 77708-862
Anti-CIDEB Rabbit Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: CIDEB
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR