Anti-CIDEB Rabbit Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A45114-50UG A45114-100UG
77708-861EA 360.84 CAD
77708-861 77708-860
Anti-CIDEB Rabbit Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: CIDEB
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR