Anti-CIDEB Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A45112-100UG A45112-50UG
77708-856EA 529.22 CAD
77708-856 77708-857
Anti-CIDEB Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: IHC:1:20-1:200

Type:
Antigen: CIDEB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR