Anti-NDUFA10 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A45072-50UL A45072-100UL
77708-787EA 360.84 CAD
77708-787 77708-786
Anti-NDUFA10 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, WB:1:500-1:2000

Type:
Antigen: NDUFA10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR