Anti-CBS Rabbit Polyclonal Antibody (Biotinylated)

Supplier: AFG BIOSCIENCE LLC
A45053-50UG A45053-100UG
77708-751EA 360.84 CAD
77708-751 77708-750
Anti-CBS Rabbit Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: CBS
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR