Anti-INTS14 Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A64454-50UG A64454-100UG
77724-055EA 360.84 CAD
77724-055 77724-054
Anti-INTS14 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: INTS14
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR