Anti-PIGS Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A64441-50UG A64441-100UG
77724-031EA 360.84 CAD
77724-031 77724-030
Anti-PIGS Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: PIGS
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR