Anti-TRIB3 Rabbit Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A64436-50UG A64436-100UG
77724-021EA 360.84 CAD
77724-021 77724-020
Anti-TRIB3 Rabbit Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: TRIB3
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR