Anti-TGS1 Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A64433-100UG A64433-50UG
77724-014EA 529.22 CAD
77724-014 77724-015
Anti-TGS1 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: TGS1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR