Anti-MTMR9 Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A64424-50UL A64424-100UL
77723-997EA 360.84 CAD
77723-997 77723-996
Anti-MTMR9 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: MTMR9
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR