Anti-MTMR9 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A64421-100UL A64421-50UL
77723-990EA 529.22 CAD
77723-990 77723-991
Anti-MTMR9 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: IF:1:50-1:200

Type:
Antigen: MTMR9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR