Anti-DIRC1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A64243-50UG A64243-100UG
77723-662EA 360.84 CAD
77723-662 77723-661
Anti-DIRC1 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: IHC:1:20-1:200

Type:
Antigen: DIRC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR