Anti-HIST1H2AC Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A64238-100UL A64238-50UL
77723-651EA 529.22 CAD
77723-651 77723-652
Anti-HIST1H2AC Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: HIST1H2AC
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR