Anti-HIST1H2AC Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A64235-50UL A64235-100UL
77723-646EA 360.84 CAD
77723-646 77723-645
Anti-HIST1H2AC Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: IHC:1:200-1:500, IF:1:50-1:200

Type:
Antigen: HIST1H2AC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR